Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNPO
Entrez Gene ID
55163
Gene Name
pyridoxamine 5'-phosphate oxidase
Gene Aliases
HEL-S-302, PDXPO
Chromosome Location
Chr.17: 47941523 - 47949308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018129.3 NP_060599.1 3 - 4 516 78
XM_005257500.3 XP_005257557.1 3 - 4 706 78
XM_011524968.2 XP_011523270.1 2 - 3 289 78
XM_017024813.1 XP_016880302.1 3 - 4 304 78
GenBank mRNA AF468030.1 - 3 - 4 363 78
AK001397.1 - 3 - 4 468 78
AK223242.1 - 3 - 4 447 78
AK303536.1 - 2 - 3 291 78
AK303665.1 - 3 - 4 468 78
BC006525.2 - 3 - 4 403 78
DA186382.1 - 3 - 4 488 78
DB228906.1 - 3 - 4 517 78
FJ224333.1 - 3 - 4 363 78

View Full Product Details