The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003128.2 | NP_003119.2 | 16 - 17 | 3814 | 82 |
NM_178313.2 | NP_842565.2 | 15 - 16 | 3911 | 82 | |
XM_005264517.2 | XP_005264574.1 | 16 - 17 | 3723 | 82 | |
XM_005264518.1 | XP_005264575.1 | 15 - 16 | 4569 | 82 | |
XM_006712087.2 | XP_006712150.1 | 16 - 17 | 3687 | 82 | |
XM_017004779.1 | XP_016860268.1 | 16 - 17 | 3830 | 82 | |
XM_017004780.1 | XP_016860269.1 | 16 - 17 | 3784 | 82 | |
XM_017004781.1 | XP_016860270.1 | 16 - 17 | 3785 | 82 | |
GenBank mRNA | AB209748.1 | - | 16 - 17 | 3692 | 82 |
AB371586.1 | - | 16 - 17 | 3804 | 82 | |
AF327441.1 | - | 15 - 16 | 3911 | 82 | |
BC137282.1 | - | 15 - 16 | 3821 | 82 | |
BC137283.1 | - | 15 - 16 | 3821 | 82 | |
EU794662.1 | - | 16 - 17 | 3814 | 82 | |
JQ599728.1 | - | 15 - 16 | 3565 | 82 | |
S65762.1 | - | 8 - 9 | 2689 | 82 |