The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128930.2 | NP_001122402.1 | 11 - 12 | 1704 | 64 |
NM_004901.4 | NP_004892.1 | 11 - 12 | 1728 | 64 | |
GenBank mRNA | AB002390.1 | - | 9 - 10 | 1258 | 64 |
AF016032.1 | - | 11 - 12 | 1633 | 64 | |
AJ131358.1 | - | 11 - 12 | 1625 | 64 | |
AK300464.1 | - | 11 - 12 | 1636 | 64 | |
BC034477.2 | - | 11 - 12 | 1668 | 64 | |
DB091432.1 | - | 3 - 4 | 448 | 64 |