Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
Chromosome Location
Chr.9: 92210207 - 92293756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002161.5 NP_002152.2 2 - 3 262 117
NM_013417.3 NP_038203.2 2 - 3 432 117
GenBank mRNA AB209234.1 - 2 - 3 171 117
AK293014.1 - 2 - 3 189 117
AK301272.1 - 2 - 3 212 117
AK308202.1 - 2 - 3 196 117
BC065552.1 - 1 - 2 115 117
BX537417.1 - 2 - 3 200 117
BX537429.1 - 2 - 3 202 117
BX647071.1 - 2 - 3 202 117
BX647236.1 - 2 - 3 194 117
BX647430.1 - 2 - 3 196 117
D28473.1 - 2 - 3 350 117
DA335066.1 - 2 - 3 262 117
U04953.1 - 2 - 3 364 117

View Full Product Details