Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGPS1
Entrez Gene ID
9453
Gene Name
geranylgeranyl diphosphate synthase 1
Gene Aliases
GGPPS, GGPPS1
Chromosome Location
Chr.1: 235328438 - 235344532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037277.1 NP_001032354.1 1 - 2 58 80
GenBank mRNA BI520491.1 - 1 - 2 61 80

View Full Product Details