Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PACS1
Entrez Gene ID
55690
Gene Name
phosphofurin acidic cluster sorting protein 1
Gene Aliases
MRD17
Chromosome Location
Chr.11: 66070276 - 66244747 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018026.3 NP_060496.2 11 - 12 1509 120
XM_011545162.1 XP_011543464.1 11 - 12 1069 120
XM_011545164.2 XP_011543466.1 11 - 12 2377 120
GenBank mRNA AB033001.1 - 4 - 5 411 120
AK056361.1 - 10 - 11 1016 120
AK096644.1 - 10 - 11 1359 120
AY320283.1 - 11 - 12 1549 120
BC010096.1 - 11 - 12 1417 120
BC052577.1 - 11 - 12 1408 120
BC055288.1 - 3 - 4 213 120

View Full Product Details