Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PACS1
Entrez Gene ID
55690
Gene Name
phosphofurin acidic cluster sorting protein 1
Gene Aliases
MRD17
Chromosome Location
Chr.11: 66070276 - 66244747 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018026.3 NP_060496.2 23 - 24 2914 73
XM_011545162.1 XP_011543464.1 23 - 24 2474 73
XM_011545164.2 XP_011543466.1 23 - 24 3782 73
GenBank mRNA AB033001.1 - 16 - 17 1816 73
AK293966.1 - 12 - 13 1525 73
AL137271.1 - 1 - 2 102 73
AY320283.1 - 23 - 24 2954 73
BC003173.1 - 8 - 9 883 73
BC009936.1 - 9 - 10 989 73
BC010096.1 - 23 - 24 2822 73
BC042909.1 - 12 - 13 1373 73
BC052577.1 - 23 - 24 2813 73
BC055288.1 - 15 - 16 1618 73

View Full Product Details