Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location
Chr.11: 13276613 - 13387268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030272.2 NP_001025443.1 5 - 6 580 92
NM_001030273.2 NP_001025444.1 6 - 7 651 92
NM_001178.5 NP_001169.3 6 - 7 635 92
NM_001297719.1 NP_001284648.1 6 - 7 635 92
NM_001297722.1 NP_001284651.1 5 - 6 580 92
NM_001297724.1 NP_001284653.1 6 - 7 644 92
XM_006718233.3 XP_006718296.1 5 - 6 522 92
XM_006718234.1 XP_006718297.1 6 - 7 588 92
XM_011520105.2 XP_011518407.2 4 - 5 686 92
XM_011520107.2 XP_011518409.2 4 - 5 681 92
XM_011520108.1 XP_011518410.1 6 - 7 628 92
XM_011520109.1 XP_011518411.1 4 - 5 383 92
XM_011520110.1 XP_011518412.1 7 - 8 643 92
XM_011520111.1 XP_011518413.1 5 - 6 458 92
XM_011520112.2 XP_011518414.1 7 - 8 643 92
XM_011520113.1 XP_011518415.1 7 - 8 643 92
XM_017017738.1 XP_016873227.1 4 - 5 684 92
XM_017017739.1 XP_016873228.1 4 - 5 680 92
XM_017017740.1 XP_016873229.1 5 - 6 522 92
XM_017017741.1 XP_016873230.1 4 - 5 383 92
XM_017017742.1 XP_016873231.1 4 - 5 383 92
XM_017017743.1 XP_016873232.1 4 - 5 383 92
XM_017017744.1 XP_016873233.1 7 - 8 643 92
XM_017017745.1 XP_016873234.1 6 - 7 588 92
XM_017017746.1 XP_016873235.1 6 - 7 598 92
XM_017017747.1 XP_016873236.1 5 - 6 458 92
XM_017017748.1 XP_016873237.1 5 - 6 515 92
GenBank mRNA AB000812.1 - 3 - 4 216 92
AB000815.1 - 4 - 5 287 92
AB000816.1 - 5 - 6 627 92
AF044288.1 - 6 - 7 545 92
AK095749.1 - 6 - 7 536 92
AK291510.1 - 5 - 6 493 92
AK315174.1 - 6 - 7 573 92
BC016674.1 - 3 - 4 179 92
BC031214.1 - 6 - 7 476 92
BC041129.1 - 6 - 7 559 92
D89722.1 - 3 - 4 192 92
U60415.1 - 5 - 6 479 92

View Full Product Details