The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019044.4 | NP_061917.3 | 20 - 21 | 1774 | 134 |
XM_011511359.1 | XP_011509661.1 | 20 - 21 | 1771 | 134 | |
XM_011511361.1 | XP_011509663.1 | 19 - 20 | 1528 | 134 | |
XM_011511362.1 | XP_011509664.1 | 19 - 20 | 1642 | 134 | |
GenBank mRNA | AB209993.1 | - | 20 - 21 | 1693 | 134 |
AK001858.1 | - | 8 - 9 | 568 | 134 | |
AK025611.1 | - | 19 - 20 | 1542 | 134 | |
AK057926.1 | - | 20 - 21 | 1635 | 134 | |
AK290722.1 | - | 20 - 21 | 1681 | 134 | |
BC028609.1 | - | 20 - 21 | 1747 | 134 | |
BC093018.1 | - | 20 - 21 | 1664 | 134 |