The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019044.4 | NP_061917.3 | 2 - 3 | 325 | 94 |
XM_006712600.2 | XP_006712663.1 | 2 - 3 | 325 | 94 | |
XM_011511359.1 | XP_011509661.1 | 2 - 3 | 325 | 94 | |
XM_011511362.1 | XP_011509664.1 | 2 - 3 | 305 | 94 | |
XM_011511363.2 | XP_011509665.1 | 2 - 3 | 325 | 94 | |
GenBank mRNA | AB209993.1 | - | 2 - 3 | 244 | 94 |
AK025611.1 | - | 1 - 2 | 93 | 94 | |
AK057926.1 | - | 2 - 3 | 186 | 94 | |
AK290722.1 | - | 2 - 3 | 232 | 94 | |
AK309040.1 | - | 2 - 3 | 232 | 94 | |
BC093018.1 | - | 2 - 3 | 215 | 94 | |
DB084596.1 | - | 2 - 3 | 325 | 94 |