Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr.9: 17906574 - 18910950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040272.5 NP_001035362.3 5 - 6 748 72
NM_052866.4 NP_443098.3 5 - 6 748 72
XM_011518063.2 XP_011516365.1 7 - 8 886 72
XM_011518064.2 XP_011516366.1 6 - 7 1001 72
XM_017015310.1 XP_016870799.1 6 - 7 7523 72
XM_017015311.1 XP_016870800.1 7 - 8 886 72
XM_017015312.1 XP_016870801.1 6 - 7 1002 72
XM_017015313.1 XP_016870802.1 6 - 7 649 72
XM_017015314.1 XP_016870803.1 7 - 8 886 72
GenBank mRNA AF176313.1 - 5 - 6 646 72
AK290667.1 - 5 - 6 734 72
AK291956.1 - 5 - 6 688 72
BC030262.1 - 5 - 6 660 72
CX758410.1 - 5 - 6 647 72

View Full Product Details