The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254749.1 | NP_001241678.1 | 1 - 2 | 387 | 66 |
NM_003617.3 | NP_003608.1 | 1 - 2 | 387 | 66 | |
GenBank mRNA | AB008109.1 | - | 1 - 2 | 130 | 66 |
AF030108.1 | - | 1 - 2 | 125 | 66 | |
AF159570.1 | - | 1 - 2 | 93 | 66 | |
AF493929.1 | - | 1 - 2 | 49 | 66 | |
AK315357.1 | - | 1 - 2 | 130 | 66 | |
AU130764.1 | - | 1 - 2 | 296 | 66 | |
BC030059.1 | - | 1 - 2 | 112 | 66 | |
BU172751.1 | - | 1 - 2 | 117 | 66 | |
BX537427.1 | - | 1 - 2 | 131 | 66 | |
DA699380.1 | - | 1 - 2 | 365 | 66 | |
DA731601.1 | - | 1 - 2 | 388 | 66 |