The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198517.3 | NP_940919.1 | 5 - 6 | 524 | 94 |
NR_046266.1 | 4 - 5 | 694 | 94 | ||
XM_006718538.3 | XP_006718601.1 | 5 - 6 | 783 | 94 | |
XM_006718539.3 | XP_006718602.1 | 5 - 6 | 766 | 94 | |
XM_006718540.3 | XP_006718603.1 | 5 - 6 | 696 | 94 | |
XM_011545002.2 | XP_011543304.1 | 4 - 5 | 744 | 94 | |
GenBank mRNA | BC022202.1 | - | 4 - 5 | 468 | 94 |
BC036873.1 | - | 4 - 5 | 479 | 94 | |
BC062999.1 | - | 5 - 6 | 496 | 94 | |
CR979868.1 | - | 5 - 6 | 524 | 94 |