The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321427.1 | NP_001308356.1 | 12 - 13 | 1521 | 78 |
NM_001321428.1 | NP_001308357.1 | 11 - 12 | 932 | 78 | |
NM_001321429.1 | NP_001308358.1 | 12 - 13 | 1108 | 78 | |
NM_001321430.1 | NP_001308359.1 | 12 - 13 | 1048 | 78 | |
NM_001321432.1 | NP_001308361.1 | 11 - 12 | 872 | 78 | |
NM_001321433.1 | NP_001308362.1 | 12 - 13 | 1045 | 78 | |
NM_001321435.1 | NP_001308364.1 | 12 - 13 | 1119 | 78 | |
NM_001321436.1 | NP_001308365.1 | 12 - 13 | 1123 | 78 | |
NM_001321438.1 | NP_001308367.1 | 10 - 11 | 882 | 78 | |
NM_022152.5 | NP_071435.2 | 11 - 12 | 947 | 78 | |
NR_135643.1 | 10 - 11 | 705 | 78 | ||
XM_011511627.1 | XP_011509929.1 | 11 - 12 | 1300 | 78 | |
GenBank mRNA | AB257295.1 | - | 11 - 12 | 1008 | 78 |
AF193045.1 | - | 11 - 12 | 861 | 78 | |
AK027755.1 | - | 9 - 10 | 595 | 78 | |
AK075465.1 | - | 7 - 8 | 422 | 78 | |
AK090618.1 | - | 12 - 13 | 1519 | 78 | |
AK093911.1 | - | 9 - 10 | 1459 | 78 | |
AK130380.1 | - | 11 - 12 | 905 | 78 | |
AK130713.1 | - | 11 - 12 | 883 | 78 | |
AK298123.1 | - | 12 - 13 | 719 | 78 | |
AK298500.1 | - | 10 - 11 | 641 | 78 | |
AK300597.1 | - | 10 - 11 | 809 | 78 | |
BC007980.2 | - | 11 - 12 | 861 | 78 | |
BC013428.1 | - | 11 - 12 | 925 | 78 | |
BC026348.1 | - | 11 - 12 | 882 | 78 | |
BC026693.2 | - | 11 - 12 | 893 | 78 | |
CA488388.1 | - | 1 - 2 | 60 | 78 |