The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321427.1 | NP_001308356.1 | 5 - 6 | 1092 | 69 |
NM_001321428.1 | NP_001308357.1 | 4 - 5 | 503 | 69 | |
NM_001321429.1 | NP_001308358.1 | 5 - 6 | 679 | 69 | |
NM_001321430.1 | NP_001308359.1 | 5 - 6 | 619 | 69 | |
NM_001321432.1 | NP_001308361.1 | 4 - 5 | 443 | 69 | |
NM_001321433.1 | NP_001308362.1 | 5 - 6 | 616 | 69 | |
NM_001321435.1 | NP_001308364.1 | 5 - 6 | 690 | 69 | |
NM_001321436.1 | NP_001308365.1 | 5 - 6 | 694 | 69 | |
NM_022152.5 | NP_071435.2 | 4 - 5 | 518 | 69 | |
NR_135643.1 | 3 - 4 | 276 | 69 | ||
XM_011511627.1 | XP_011509929.1 | 4 - 5 | 871 | 69 | |
GenBank mRNA | AB257295.1 | - | 4 - 5 | 579 | 69 |
AF193045.1 | - | 4 - 5 | 432 | 69 | |
AK027755.1 | - | 2 - 3 | 166 | 69 | |
AK090618.1 | - | 5 - 6 | 1090 | 69 | |
AK093911.1 | - | 2 - 3 | 1030 | 69 | |
AK130380.1 | - | 4 - 5 | 476 | 69 | |
AK130713.1 | - | 4 - 5 | 454 | 69 | |
AK298123.1 | - | 5 - 6 | 290 | 69 | |
AK298500.1 | - | 3 - 4 | 212 | 69 | |
BC007980.2 | - | 4 - 5 | 432 | 69 | |
BC013428.1 | - | 4 - 5 | 496 | 69 | |
BC026348.1 | - | 4 - 5 | 453 | 69 | |
BC026693.2 | - | 4 - 5 | 464 | 69 | |
CF994144.1 | - | 4 - 5 | 480 | 69 |