The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167890.1 | NP_001161362.1 | 7 - 8 | 1064 | 73 |
NM_015507.3 | NP_056322.2 | 7 - 8 | 1064 | 73 | |
GenBank mRNA | AF186084.1 | - | 7 - 8 | 922 | 73 |
AF193055.1 | - | 7 - 8 | 989 | 73 | |
AJ245671.1 | - | 7 - 8 | 1017 | 73 | |
AK075214.1 | - | 7 - 8 | 1034 | 73 | |
AK300137.1 | - | 7 - 8 | 800 | 73 | |
AK315018.1 | - | 7 - 8 | 902 | 73 | |
AL117610.1 | - | 7 - 8 | 794 | 73 | |
AY358333.1 | - | 7 - 8 | 911 | 73 | |
BC038587.1 | - | 7 - 8 | 873 | 73 |