Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SH3KBP1
- Entrez Gene ID
-
30011
- Gene Name
- SH3 domain containing kinase binding protein 1
- Gene Aliases
-
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
- Chromosome Location
-
Chr.X: 19533965 - 19887626 on Build GRCh38
View Full Product Details