The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014901.4 | NP_055716.1 | 6 - 7 | 1325 | 76 |
XM_005265840.2 | XP_005265897.1 | 6 - 7 | 1037 | 76 | |
XM_005265841.4 | XP_005265898.1 | 6 - 7 | 1282 | 76 | |
XM_005265842.4 | XP_005265899.1 | 6 - 7 | 1267 | 76 | |
XM_005265843.2 | XP_005265900.1 | 6 - 7 | 973 | 76 | |
XM_005265844.3 | XP_005265901.1 | 6 - 7 | 1078 | 76 | |
XM_005265845.4 | XP_005265902.1 | 6 - 7 | 1216 | 76 | |
XM_006714831.2 | XP_006714894.1 | 6 - 7 | 1028 | 76 | |
XM_006714832.2 | XP_006714895.1 | 6 - 7 | 896 | 76 | |
XM_011534466.2 | XP_011532768.1 | 6 - 7 | 1040 | 76 | |
XM_011534467.2 | XP_011532769.1 | 6 - 7 | 1271 | 76 | |
GenBank mRNA | AB029023.1 | - | 6 - 7 | 1210 | 76 |
AK302385.1 | - | 6 - 7 | 1078 | 76 | |
BC039833.1 | - | 6 - 7 | 1281 | 76 | |
BC063297.1 | - | 6 - 7 | 1326 | 76 |