Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELL2
Entrez Gene ID
4753
Gene Name
neural EGFL like 2
Gene Aliases
NRP2
Chromosome Location
Chr.12: 44508275 - 44921848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145107.1 NP_001138579.1 11 - 12 1362 112
NM_001145108.1 NP_001138580.1 10 - 11 1682 112
NM_001145109.1 NP_001138581.1 10 - 11 1187 112
NM_001145110.1 NP_001138582.1 11 - 12 1248 112
NM_006159.2 NP_006150.1 11 - 12 1289 112
XM_005268905.3 XP_005268962.1 11 - 12 1442 112
XM_011538396.1 XP_011536698.1 12 - 13 1238 112
XM_017019341.1 XP_016874830.1 11 - 12 1618 112
XM_017019342.1 XP_016874831.1 11 - 12 1301 112
XM_017019343.1 XP_016874832.1 11 - 12 1457 112
XM_017019344.1 XP_016874833.1 12 - 13 1253 112
GenBank mRNA AK056082.1 - 10 - 11 1533 112
AK095637.1 - 11 - 12 1673 112
AK295125.1 - 11 - 12 1248 112
AK297521.1 - 12 - 13 1277 112
AK299277.1 - 10 - 11 1143 112
AK299733.1 - 11 - 12 112
AK309825.1 - 12 - 13 1273 112
AK315960.1 - 10 - 11 1116 112
AK316058.1 - 11 - 12 1609 112
BC020544.1 - 10 - 11 1389 112
D83018.1 - 11 - 12 1183 112
D89629.1 - 10 - 11 1084 112

View Full Product Details