The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015483.2 | NP_056298.2 | 1 - 2 | 323 | 87 |
XM_005249697.2 | XP_005249754.1 | 1 - 2 | 178 | 87 | |
GenBank mRNA | AK292802.1 | - | 1 - 2 | 45 | 87 |
BC037887.2 | - | 1 - 2 | 227 | 87 | |
BC047107.1 | - | 1 - 2 | 60 | 87 |