The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015483.2 | NP_056298.2 | 3 - 4 | 988 | 139 |
XM_005249696.2 | XP_005249753.1 | 3 - 4 | 844 | 139 | |
XM_005249697.2 | XP_005249754.1 | 3 - 4 | 577 | 139 | |
XM_011515256.2 | XP_011513558.1 | 3 - 4 | 723 | 139 | |
XM_011515257.2 | XP_011513559.1 | 3 - 4 | 457 | 139 | |
GenBank mRNA | AF151831.1 | - | 2 - 3 | 397 | 139 |
AK292802.1 | - | 3 - 4 | 710 | 139 | |
BC037887.2 | - | 3 - 4 | 892 | 139 | |
BC047107.1 | - | 3 - 4 | 725 | 139 |