The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032861.3 | NP_116250.3 | 4 - 5 | 409 | 159 |
NR_073096.1 | 4 - 5 | 409 | 159 | ||
XM_006715586.2 | XP_006715649.1 | 3 - 4 | 338 | 159 | |
XM_011536196.2 | XP_011534498.1 | 3 - 4 | 432 | 159 | |
XM_011536198.2 | XP_011534500.1 | 2 - 3 | 334 | 159 | |
GenBank mRNA | AK027823.1 | - | 4 - 5 | 399 | 159 |
AK125878.1 | - | 4 - 5 | 1501 | 159 | |
BC001705.1 | - | 4 - 5 | 366 | 159 | |
BC028594.1 | - | 5 - 6 | 3608 | 159 | |
BX648104.1 | - | 3 - 4 | 341 | 159 | |
DA232092.1 | - | 4 - 5 | 409 | 159 |