The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032861.3 | NP_116250.3 | 6 - 7 | 626 | 74 |
NR_073096.1 | 6 - 7 | 626 | 74 | ||
XM_006715586.2 | XP_006715649.1 | 5 - 6 | 555 | 74 | |
XM_011536196.2 | XP_011534498.1 | 5 - 6 | 649 | 74 | |
XM_011536198.2 | XP_011534500.1 | 4 - 5 | 551 | 74 | |
GenBank mRNA | AK027823.1 | - | 6 - 7 | 616 | 74 |
BC001705.1 | - | 6 - 7 | 583 | 74 | |
BX648104.1 | - | 5 - 6 | 558 | 74 | |
DB046171.1 | - | 5 - 6 | 500 | 74 |