The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146070.1 | NP_001139542.1 | 9 - 10 | 1087 | 112 |
NM_001146071.1 | NP_001139543.1 | 9 - 10 | 1524 | 112 | |
NM_030794.2 | NP_110421.1 | 9 - 10 | 1524 | 112 | |
XM_005266556.4 | XP_005266613.1 | 9 - 10 | 1375 | 112 | |
XM_005266560.2 | XP_005266617.1 | 9 - 10 | 1482 | 112 | |
XM_011535247.1 | XP_011533549.1 | 8 - 9 | 974 | 112 | |
XM_011535249.2 | XP_011533551.1 | 8 - 9 | 974 | 112 | |
XM_017020777.1 | XP_016876266.1 | 9 - 10 | 1375 | 112 | |
GenBank mRNA | AK024660.1 | - | 9 - 10 | 993 | 112 |
BC030514.1 | - | 9 - 10 | 1046 | 112 | |
BC060876.1 | - | 9 - 10 | 1011 | 112 | |
BX537910.1 | - | 9 - 10 | 1524 | 112 | |
EU643838.1 | - | 9 - 10 | 1087 | 112 |