Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOM2
Entrez Gene ID
9172
Gene Name
myomesin 2
Gene Aliases
TTNAP
Chromosome Location
Chr.8: 2045043 - 2145456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003970.3 NP_003961.3 12 - 13 1599 66
XM_006716237.1 XP_006716300.1 12 - 13 1564 66
GenBank mRNA BC052969.1 - 12 - 13 1552 66
X69089.1 - 12 - 13 1509 66

View Full Product Details