The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003970.3 | NP_003961.3 | 29 - 30 | 3698 | 120 |
XM_006716237.1 | XP_006716300.1 | 29 - 30 | 3663 | 120 | |
GenBank mRNA | AK297890.1 | - | 16 - 17 | 2016 | 120 |
BC052969.1 | - | 29 - 30 | 3651 | 120 | |
BP260135.1 | - | 4 - 5 | 371 | 120 | |
BP260324.1 | - | 4 - 5 | 375 | 120 | |
X69089.1 | - | 29 - 30 | 3608 | 120 |