Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
Chromosome Location
Chr.8: 102826302 - 102864200 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301668.1 NP_001288597.1 6 - 7 1212 101
NM_015878.5 NP_056962.2 7 - 8 1358 101
NM_148174.3 NP_680479.1 6 - 7 1212 101
XM_005250969.1 XP_005251026.1 6 - 7 1173 101
XM_011517126.1 XP_011515428.1 7 - 8 1345 101
XM_011517127.1 XP_011515429.1 7 - 8 1336 101
XM_011517128.2 XP_011515430.1 6 - 7 1356 101
XM_017013563.1 XP_016869052.1 7 - 8 1324 101
XM_017013564.1 XP_016869053.1 7 - 8 1336 101
XM_017013565.1 XP_016869054.1 7 - 8 1475 101
XM_017013566.1 XP_016869055.1 6 - 7 1356 101
GenBank mRNA AK095493.1 - 6 - 7 1140 101
AK290516.1 - 7 - 8 1090 101
BC013420.2 - 7 - 8 1297 101
BC019279.1 - 6 - 7 1159 101
BC042404.1 - 6 - 7 1183 101
BT006858.1 - 4 - 5 585 101
CR456745.1 - 4 - 5 585 101
D88674.1 - 7 - 8 1294 101

View Full Product Details