Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TCF7
- Entrez Gene ID
-
6932
- Gene Name
- transcription factor 7 (T-cell specific, HMG-box)
- Gene Aliases
-
TCF-1
- Chromosome Location
-
Chr.5: 134114703 - 134148229 on Build GRCh38
View Full Product Details