The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011512208.1 | XP_011510510.1 | 1 - 2 | 303 | 151 |
XM_011512212.1 | XP_011510514.1 | 1 - 2 | 302 | 151 | |
XM_017005382.1 | XP_016860871.1 | 1 - 2 | 302 | 151 |