The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004278.3 | NP_004269.1 | 4 - 5 | 524 | 75 |
XM_017025352.1 | XP_016880841.1 | 4 - 5 | 510 | 75 | |
XM_017025353.1 | XP_016880842.1 | 4 - 5 | 510 | 75 | |
GenBank mRNA | AB017165.1 | - | 4 - 5 | 497 | 75 |
AK292932.1 | - | 4 - 5 | 509 | 75 | |
BC068197.1 | - | 4 - 5 | 503 | 75 |