The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000926.4 | NP_000917.3 | 3 - 4 | 2647 | 128 |
NM_001202474.3 | NP_001189403.1 | 3 - 4 | 1897 | 128 | |
NM_001271162.1 | NP_001258091.1 | 3 - 4 | 567 | 128 | |
NR_073141.2 | 3 - 4 | 1897 | 128 | ||
XM_006718858.3 | XP_006718921.1 | 3 - 4 | 3497 | 128 | |
GenBank mRNA | AB085845.1 | - | 3 - 4 | 1904 | 128 |
AF016381.1 | - | 3 - 4 | 2079 | 128 | |
AK304853.1 | - | 3 - 4 | 567 | 128 | |
AY382151.1 | - | 2 - 3 | 263 | 128 | |
M15716.1 | - | 3 - 4 | 2079 | 128 | |
X51730.1 | - | 3 - 4 | 3358 | 128 |