The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018351.3 | NP_060821.3 | 20 - 21 | 4477 | 175 |
GenBank mRNA | AB037783.1 | - | 19 - 20 | 2248 | 175 |
AK002045.1 | - | 13 - 14 | 1268 | 175 | |
AK124904.1 | - | 20 - 21 | 4477 | 175 | |
AK128492.1 | - | 19 - 20 | 3354 | 175 | |
BC125225.1 | - | 19 - 20 | 1880 | 175 | |
BC125226.1 | - | 19 - 20 | 1880 | 175 |