Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD6
Entrez Gene ID
55785
Gene Name
FYVE, RhoGEF and PH domain containing 6
Gene Aliases
ZFYVE24
Chromosome Location
Chr.12: 95076749 - 95217464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018351.3 NP_060821.3 14 - 15 3722 103
GenBank mRNA AB037783.1 - 13 - 14 1495 103
AK002045.1 - 7 - 8 513 103
AK124904.1 - 14 - 15 3722 103
AK128492.1 - 13 - 14 2599 103
BC013319.1 - 8 - 9 605 103
BC125225.1 - 13 - 14 1125 103
BC125226.1 - 13 - 14 1125 103
BX538322.1 - 14 - 15 3638 103

View Full Product Details