Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
Chromosome Location
Chr.1: 109483904 - 109492804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153340.4 NP_699171.3 3 - 4 309 77
XM_005270445.3 XP_005270502.1 3 - 4 344 77
XM_005270446.2 XP_005270503.1 3 - 4 340 77
XM_011540638.2 XP_011538940.1 2 - 3 243 77
XM_011540639.2 XP_011538941.1 3 - 4 1613 77
XM_011540641.2 XP_011538943.1 3 - 4 946 77
XM_011540644.2 XP_011538946.1 5 - 6 428 77
XM_011540645.1 XP_011538947.1 3 - 4 272 77
XM_011540646.2 XP_011538948.1 3 - 4 337 77
XM_017000267.1 XP_016855756.1 4 - 5 1075 77
XM_017000268.1 XP_016855757.1 4 - 5 408 77
XM_017000269.1 XP_016855758.1 4 - 5 1075 77
XM_017000270.1 XP_016855759.1 4 - 5 476 77
GenBank mRNA AK090460.1 - 1 - 2 1061 77

View Full Product Details