The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152434.2 | NP_689647.2 | 3 - 4 | 361 | 161 |
XM_011542620.2 | XP_011540922.1 | 3 - 4 | 359 | 161 | |
XM_017017239.1 | XP_016872728.1 | 3 - 4 | 332 | 161 | |
GenBank mRNA | AK126881.1 | - | 3 - 4 | 354 | 161 |
AK293001.1 | - | 3 - 4 | 296 | 162 | |
BC070142.1 | - | 3 - 4 | 161 | ||
BC107418.1 | - | 3 - 4 | 344 | 162 | |
BX647832.1 | - | 3 - 4 | 335 | 161 | |
BX648704.1 | - | 3 - 4 | 340 | 161 | |
BX649022.1 | - | 3 - 4 | 331 | 161 | |
DA291472.1 | - | 3 - 4 | 363 | 161 |