The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007011.7 | NP_008942.3 | 7 - 8 | 1118 | 86 |
NM_152924.4 | NP_690888.1 | 3 - 4 | 720 | 86 | |
XM_005254832.1 | XP_005254889.1 | 6 - 7 | 756 | 86 | |
XM_005254833.1 | XP_005254890.1 | 3 - 4 | 364 | 86 | |
XM_011521165.1 | XP_011519467.1 | 4 - 5 | 569 | 86 | |
GenBank mRNA | AF546700.1 | - | 3 - 4 | 452 | 86 |
AK222841.1 | - | 3 - 4 | 392 | 86 | |
AK223083.1 | - | 3 - 4 | 390 | 86 | |
AK290340.1 | - | 3 - 4 | 392 | 86 | |
BC019248.1 | - | 7 - 8 | 848 | 86 | |
BC090052.1 | - | 3 - 4 | 454 | 86 | |
BI464471.1 | - | 3 - 4 | 485 | 86 | |
X12433.1 | - | 3 - 4 | 375 | 86 |