Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO1C
Entrez Gene ID
4641
Gene Name
myosin IC
Gene Aliases
MMI-beta, MMIb, NMI, myr2
Chromosome Location
Chr.17: 1464186 - 1492707 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080779.1 NP_001074248.1 31 - 32 3391 73
NM_001080950.1 NP_001074419.1 31 - 32 3160 73
NM_033375.4 NP_203693.3 31 - 32 3175 73
GenBank mRNA AB210015.1 - 31 - 32 3341 73
AK172742.1 - 12 - 13 1635 73
AK292910.1 - 31 - 32 3370 73
AK295761.1 - 31 - 32 3330 73
BC044891.1 - 30 - 31 3072 73
BC068013.1 - 31 - 32 3385 73

View Full Product Details