The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080779.1 | NP_001074248.1 | 3 - 4 | 570 | 69 |
NM_001080950.1 | NP_001074419.1 | 3 - 4 | 339 | 69 | |
NM_033375.4 | NP_203693.3 | 3 - 4 | 354 | 69 | |
GenBank mRNA | AB210015.1 | - | 3 - 4 | 520 | 69 |
AK172742.1 | - | 3 - 4 | 551 | 69 | |
AK292910.1 | - | 3 - 4 | 549 | 69 | |
AK295761.1 | - | 3 - 4 | 509 | 69 | |
AK304782.1 | - | 3 - 4 | 397 | 69 | |
BC044891.1 | - | 2 - 3 | 251 | 69 | |
BC068013.1 | - | 3 - 4 | 564 | 69 | |
BP255642.1 | - | 3 - 4 | 354 | 69 | |
BP284965.1 | - | 3 - 4 | 334 | 69 | |
CD050346.1 | - | 3 - 4 | 596 | 69 | |
X98507.1 | - | 2 - 3 | 310 | 69 |