Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB11
Entrez Gene ID
51726
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Gene Aliases
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
Chromosome Location
Chr.3: 186570676 - 186585800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016306.5 NP_057390.1 1 - 2 290 126
GenBank mRNA AB028859.2 - 1 - 2 233 126
AF228505.1 - 1 - 2 159 126
AF277317.1 - 1 - 2 250 126
AJ250137.1 - 1 - 2 88 126
AK075300.1 - 1 - 2 247 126
AK075430.1 - 1 - 2 266 126
AY359043.1 - 1 - 2 226 126
BC001144.1 - 1 - 2 228 126
BC046500.1 - 2 - 3 997 126
BM754506.1 - 1 - 2 277 126
BT007063.1 - 1 - 2 68 126
CR457096.1 - 1 - 2 68 126
DA058195.1 - 1 - 2 290 126

View Full Product Details