Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC27A5
Entrez Gene ID
10998
Gene Name
solute carrier family 27 member 5
Gene Aliases
ACSB, ACSVL6, BACS, BAL, FACVL3, FATP-5, FATP5, VLACSR, VLCS-H2, VLCSH2
Chromosome Location
Chr.19: 58498333 - 58512065 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321196.1 NP_001308125.1 4 - 5 1037 65
NM_012254.2 NP_036386.1 4 - 5 1289 65
XM_011526364.1 XP_011524666.1 4 - 5 1218 65
GenBank mRNA AF064255.1 - 4 - 5 1249 65
AK123036.1 - 4 - 5 1289 65
AK298446.1 - 4 - 5 964 65

View Full Product Details