Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC27A5
Entrez Gene ID
10998
Gene Name
solute carrier family 27 member 5
Gene Aliases
ACSB, ACSVL6, BACS, BAL, FACVL3, FATP-5, FATP5, VLACSR, VLCS-H2, VLCSH2
Chromosome Location
Chr.19: 58498333 - 58512065 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321196.1 NP_001308125.1 8 - 9 1623 64
NM_012254.2 NP_036386.1 8 - 9 1875 64
GenBank mRNA AF064255.1 - 8 - 9 1835 64
AK026640.1 - 1 - 2 141 64
AK123036.1 - 8 - 9 1875 64
AK298446.1 - 8 - 9 1550 64

View Full Product Details