The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271754.1 | NP_001258683.1 | 4 - 5 | 747 | 78 |
NM_001271755.1 | NP_001258684.1 | 4 - 5 | 818 | 78 | |
NM_001271756.1 | NP_001258685.1 | 4 - 5 | 457 | 78 | |
NM_004791.2 | NP_004782.1 | 5 - 6 | 965 | 78 | |
XM_005254100.4 | XP_005254157.1 | 5 - 6 | 923 | 78 | |
GenBank mRNA | AB008375.1 | - | 6 - 7 | 829 | 78 |
AF072752.1 | - | 5 - 6 | 943 | 78 | |
AK095875.1 | - | 5 - 6 | 605 | 78 | |
AK291340.1 | - | 5 - 6 | 885 | 78 | |
AK297744.1 | - | 4 - 5 | 673 | 78 | |
AK298571.1 | - | 4 - 5 | 457 | 78 | |
AK308194.1 | - | 4 - 5 | 735 | 78 | |
BC036788.1 | - | 5 - 6 | 886 | 78 |