The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003291.2 | NP_003282.2 | 11 - 12 | 1493 | 119 |
XM_005254070.1 | XP_005254127.1 | 11 - 12 | 1492 | 119 | |
XM_005254071.1 | XP_005254128.1 | 11 - 12 | 1492 | 119 | |
XM_005254072.1 | XP_005254129.1 | 11 - 12 | 1492 | 119 | |
GenBank mRNA | AK304313.1 | - | 11 - 12 | 1401 | 119 |
BC039905.1 | - | 12 - 13 | 1436 | 119 | |
M55169.1 | - | 11 - 12 | 1225 | 119 | |
M73047.1 | - | 11 - 12 | 1413 | 119 |