Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCEL
Entrez Gene ID
8796
Gene Name
sciellin
Gene Aliases
-
Chromosome Location
Chr.13: 77535674 - 77645263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160706.1 NP_001154178.1 13 - 14 962 96
NM_003843.3 NP_003834.3 13 - 14 968 96
NM_144777.2 NP_659001.2 14 - 15 1028 96
XM_005266578.1 XP_005266635.1 13 - 14 939 96
XM_006719884.1 XP_006719947.1 13 - 14 908 96
XM_011535281.1 XP_011533583.1 14 - 15 974 96
XM_011535282.1 XP_011533584.1 14 - 15 974 96
XM_011535283.1 XP_011533585.1 14 - 15 974 96
XM_011535284.1 XP_011533586.1 14 - 15 974 96
XM_011535285.1 XP_011533587.1 13 - 14 984 96
XM_011535286.1 XP_011533588.1 14 - 15 974 96
XM_011535287.1 XP_011533589.1 14 - 15 974 96
XM_011535288.1 XP_011533590.1 12 - 13 838 96
XM_011535290.1 XP_011533592.1 14 - 15 974 96
XM_011535291.1 XP_011533593.1 14 - 15 974 96
XM_017020805.1 XP_016876294.1 14 - 15 974 96
GenBank mRNA AF045941.1 - 13 - 14 884 96
AK025320.1 - 14 - 15 942 96
AK222720.1 - 14 - 15 942 96
AK301659.1 - 13 - 14 876 96
BC047536.1 - 13 - 14 1043 96

View Full Product Details