The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160706.1 | NP_001154178.1 | 26 - 27 | 1737 | 110 |
NM_003843.3 | NP_003834.3 | 27 - 28 | 1803 | 110 | |
NM_144777.2 | NP_659001.2 | 28 - 29 | 1863 | 110 | |
XM_005266578.1 | XP_005266635.1 | 27 - 28 | 1774 | 110 | |
XM_006719882.1 | XP_006719945.1 | 27 - 28 | 1749 | 110 | |
XM_006719884.1 | XP_006719947.1 | 27 - 28 | 1743 | 110 | |
XM_006719885.1 | XP_006719948.1 | 26 - 27 | 1689 | 110 | |
XM_011535281.1 | XP_011533583.1 | 28 - 29 | 1806 | 110 | |
XM_011535282.1 | XP_011533584.1 | 28 - 29 | 1806 | 110 | |
XM_011535283.1 | XP_011533585.1 | 28 - 29 | 1806 | 110 | |
XM_011535284.1 | XP_011533586.1 | 28 - 29 | 1803 | 110 | |
XM_011535285.1 | XP_011533587.1 | 27 - 28 | 1819 | 110 | |
XM_011535286.1 | XP_011533588.1 | 27 - 28 | 1749 | 110 | |
XM_011535287.1 | XP_011533589.1 | 27 - 28 | 1746 | 110 | |
XM_011535288.1 | XP_011533590.1 | 26 - 27 | 1673 | 110 | |
XM_011535289.1 | XP_011533591.1 | 26 - 27 | 1686 | 110 | |
GenBank mRNA | AF045941.1 | - | 27 - 28 | 1719 | 110 |
AK025320.1 | - | 28 - 29 | 1777 | 110 | |
AK222720.1 | - | 28 - 29 | 1777 | 110 | |
AK301659.1 | - | 26 - 27 | 1651 | 110 | |
BC047536.1 | - | 27 - 28 | 1878 | 110 |