The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130916.2 | NP_001124388.1 | 1 - 2 | 214 | 139 |
NM_001306210.1 | NP_001293139.1 | 1 - 2 | 214 | 139 | |
NM_004612.3 | NP_004603.1 | 1 - 2 | 214 | 139 | |
GenBank mRNA | AJ619019.2 | - | 1 - 2 | 173 | 139 |
AK302059.1 | - | 1 - 2 | 214 | 139 | |
BC071181.1 | - | 1 - 2 | 192 | 139 | |
L11695.1 | - | 1 - 2 | 173 | 139 |