The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001855.4 | NP_001846.3 | 13 - 14 | 2106 | 130 |
XM_011518214.2 | XP_011516516.1 | 14 - 15 | 2101 | 130 | |
GenBank mRNA | AK095885.1 | - | 13 - 14 | 1822 | 130 |
L01697.1 | - | 3 - 4 | 128 | 130 | |
L25286.1 | - | 13 - 14 | 1846 | 130 |