The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001855.4 | NP_001846.3 | 17 - 18 | 2431 | 58 |
XM_011518214.2 | XP_011516516.1 | 18 - 19 | 2426 | 58 | |
GenBank mRNA | AK095885.1 | - | 17 - 18 | 2147 | 58 |
L01697.1 | - | 7 - 8 | 453 | 58 | |
L25286.1 | - | 17 - 18 | 2171 | 58 |