The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042450.2 | NP_001035915.1 | 7 - 8 | 677 | 75 |
NM_001270649.1 | NP_001257578.1 | 7 - 8 | 677 | 75 | |
NM_152351.4 | NP_689564.3 | 7 - 8 | 677 | 75 | |
XM_011523652.1 | XP_011521954.1 | 7 - 8 | 833 | 75 | |
XM_017024191.1 | XP_016879680.1 | 7 - 8 | 834 | 75 | |
GenBank mRNA | AK057946.1 | - | 7 - 8 | 678 | 75 |
AK298345.1 | - | 7 - 8 | 663 | 75 | |
BC034380.1 | - | 7 - 8 | 622 | 75 |