Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC19
Entrez Gene ID
64922
Gene Name
leucine rich repeat containing 19
Gene Aliases
-
Chromosome Location
Chr.9: 26993136 - 27005693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022901.2 NP_075052.1 2 - 3 197 124
GenBank mRNA AK024955.1 - 2 - 3 176 124
AK222987.1 - 2 - 3 169 124
BC056252.1 - 2 - 3 201 124
BC126156.1 - 2 - 3 156 124

View Full Product Details